Learn More
Medchemexpress LLC HY-D1056 5mg Medchemexpress, Lipopolysaccharides from Escherichia coli O55:B5 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD10565MG
Encompass_Preferred
Medchemexpress, HY-D1056 5mg Lipopolysaccharides from Escherichia coli O55:B5 CAS: Lipopolysaccharides from Escherichia coli O55:B5 (LPS) is an endotoxin derived from the outer leaflet of the outer membrane of Gram-negative bacteria. Lipopolysaccharides from Escherichia coli O55:B5 consists of an antigen O-specific chain, a core oligosaccharide and lipid A[3]. Lipopolysaccharides from Escherichia coli O55:B5 is a highly immunogenic antigen which can enhance immune responses[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0197