Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from E10MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L302410MG
Encompass
This product is extracted from E. coli serotype O111B4 and purified by ion exchange. The source strain is from a private collection. This LPS serotype has been used to stimulate B-cells and induce NOS in human hepatocytes.
Catalog No. 50-290-4325