Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from E10MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L452410MG
Encompass
This product is extracted from E. coli serotype O55B5 and purified by ion exchange. The source strain is CDC 1644-70. The LPS O55B5 has been used to stimulate human peritoneal macrophages at 1 ng/mL and to stimulate equine peritoneal macrophages at 1-100 ng/mL.
Catalog No. 50-290-4817