Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from E10MG
Catalog No. 502904142
Encompass
This product is extracted from E. coli serotype O55B5 and purified by gel filtration. The source strain is CDC 1644-70. The LPS O55B5 has been used to stimulate human peritoneal macrophages at 1 ng/ml and to stimulate equine peritoneal macrophages at 1-100 ng/ml.
Catalog No. 50-290-4142 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L263710MG
This product is extracted from E. coli serotype O55B5 and purified by gel filtration. The source strain is CDC 1644-70. The LPS O55B5 has been used to stimulate human peritoneal macrophages at 1 ng/ml and to stimulate equine peritoneal macrophages at 1-100 ng/ml.