Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from E25MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L502425MG
Encompass
This product is extracted from E. coli serotype O127B8 and purified by ion exchange. The source strain is ATCC 12740. This LPS serotype has been used in the study of septic shock and to induce NOS in murine macrophages and PAf synthesis in rat glomerular masangial cells.
Catalog No. 50-291-4423