Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from Escherichia coli O26:B6, MFCD00164401, 1 mg
Catalog No. NC0736547
Encompass
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs). Lipopolysaccharides from Escherichia coli 026:B6 was used to elicit the secretion of cytokines by human PBMC, murine bone marrow dendritic cells and rat astrocytes.
Catalog No. NC0736547 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L26541MG
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs). Lipopolysaccharides from Escherichia coli 026:B6 was used to elicit the secretion of cytokines by human PBMC, murine bone marrow dendritic cells and rat astrocytes.