Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from K10MG
Catalog No. 502900014
Encompass
This product is phenol extracted from Klebsiella pneumoniae. The source strain is ATCC 15380.
Catalog No. 50-290-0014 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L426810MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This product is phenol extracted from Klebsiella pneumoniae. The source strain is ATCC 15380.