Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from P10MG
Catalog No. 502900023
Encompass
This product is phenol extracted from Pseudomonas aeruginosa serotype 10.22 The source strain is ATCC 27316.
Catalog No. 50-290-0023 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L914310MG
This product is phenol extracted from Pseudomonas aeruginosa serotype 10.22 The source strain is ATCC 27316.