Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from S25MG
Catalog No. 502900124
Encompass
This product is phenol extracted from Salmonella typhimurium. The source strain is ATCC 7823. This LPS has been used to induce NOS in rats and guinea pigs.
Catalog No. 50-290-0124 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L651125MG
This product is phenol extracted from Salmonella typhimurium. The source strain is ATCC 7823. This LPS has been used to induce NOS in rats and guinea pigs.