Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lipopolysaccharides, from Klebsiella pneumoniae | 10-100 kDa | 10 MG
SDP

Catalog No. 5000297177
Encompass_Preferred

Lipopolysaccharides isolated from Klebsiella pneumoniae are bacterial endotoxins that activate Toll-like receptor 4 (TLR4) and are used to model inflammation in immunology research. Supplied as a powder with available quantity options and supporting documentation for purity and handling.

  • Source organism: Klebsiella pneumoniae
  • Application: TLR4 activation and inflammation research
  • Form: powder for dissolution
  • Molecular weight: heterogeneous, typically 10-100 kDa
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Documentation: purity and supporting documents available

Catalog No. 50-002-97177 Supplier Medchemexpress LLC Supplier No. HYD1056B310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lipopolysaccharides isolated from Klebsiella pneumoniae are bacterial endotoxins that activate Toll-like receptor 4 (TLR4) and are used to model inflammation in immunology research. Supplied as a powder with available quantity options and supporting documentation for purity and handling.

  • Source organism: Klebsiella pneumoniae
  • Application: TLR4 activation and inflammation research
  • Form: powder for dissolution
  • Molecular weight: heterogeneous, typically 10-100 kDa
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Documentation: purity and supporting documents available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.