Learn More
List Biological Laboratories Inc LPS from Escherichia coli O55:B5, 2.5 mg Lyophilized Powder
SDP
Catalog No. 501870964
Encompass_Preferred
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development projects! LPS is one of the most potent natural products capable of pluripotential immunostimulation manifested by the activation of host cells (e.g. B lymphocytes, macrophages) to functional differentiation. Host cell activation by LPS produces a spectrum of hormone-active lymphokines and monokines, including interferons (α, ß, γ), IL-1 and -6, TNF, PAF and procoagulant tissue factor. The documented capacity of LPS or their active lipid A component to initiate a variety of biochemical pathways (protein kinase C, cAMP dependent protein kinase, phosphatidyl inositol turnover, arachidonate metabolism, protein myristolation and activation of G-proteins) provides investigators with powerful molecular tools by which to study cellular activation mechanisms.
Catalog No. 50-187-0964 Supplier List Biological Laboratories Inc Supplier No. 203A
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development projects! LPS is one of the most potent natural products capable of pluripotential immunostimulation manifested by the activation of host cells (e.g. B lymphocytes, macrophages) to functional differentiation. Host cell activation by LPS produces a spectrum of hormone-active lymphokines and monokines, including interferons (α, ß, γ), IL-1 and -6, TNF, PAF and procoagulant tissue factor. The documented capacity of LPS or their active lipid A component to initiate a variety of biochemical pathways (protein kinase C, cAMP dependent protein kinase, phosphatidyl inositol turnover, arachidonate metabolism, protein myristolation and activation of G-proteins) provides investigators with powerful molecular tools by which to study cellular activation mechanisms.