Learn More
Medchemexpress LLC Lipopolysaccharides, from P. gingivalis | 1 MG
SDP
Catalog No. 5000428552
Encompass_Preferred
Lipopolysaccharides, from P. gingivalis (LPS) are endotoxins and TLR4 activators extracted from *Porphyromonas gingivalis*. This product possesses a typical three-part structure: O-antigen, core oligosaccharide, and lipid A. It activates TLR-4 in immune cells and is an important virulence factor in the mechanism of periodontal disease. It can be used in research related to periodontitis.
  • Extracted from *Porphyromonas gingivalis*
  • Classified as S (smooth) type LPS
  • Possesses O-antigen, core oligosaccharide, and lipid A structure
  • Activates TLR-4 in immune cells
  • Important virulence factor in periodontal disease
  • Suitable for periodontitis research
Catalog No. 50-004-28552 Supplier Medchemexpress LLC Supplier No. HYD1056D1MG
Lipopolysaccharides, from P. gingivalis (LPS) are endotoxins and TLR4 activators extracted from *Porphyromonas gingivalis*. This product possesses a typical three-part structure: O-antigen, core oligosaccharide, and lipid A. It activates TLR-4 in immune cells and is an important virulence factor in the mechanism of periodontal disease. It can be used in research related to periodontitis.
  • Extracted from *Porphyromonas gingivalis*
  • Classified as S (smooth) type LPS
  • Possesses O-antigen, core oligosaccharide, and lipid A structure
  • Activates TLR-4 in immune cells
  • Important virulence factor in periodontal disease
  • Suitable for periodontitis research