Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides rough10MG
Supplier: Sigma Aldrich Fine Chemicals Biosciences L976410MG
This product is prepared from Salmonella Minnesota strain Re 595 (Re mutant) using a phenol-chloroform-petroleum ether extraction procedure. The source strain is ATCC 49284. A unique structure in the LPS of strain Re 595 was shown to induce secretion and aggregation in human platelets.