Learn More
Medchemexpress LLC Lipopolysaccharides from Salmonella typhosa | 10 MG
SDP
Catalog No. 5000297134
Encompass_Preferred
Lipopolysaccharides from Salmonella typhosa are complex bacterial endotoxins composed of lipid A, a core oligosaccharide, and an O-antigen polysaccharide. Supplied as a white to off-white powder for research use, they act as Toll-like receptor 4 (TLR-4) agonists and can be used as immunostimulatory reagents or vaccine adjuvants. Storage recommendations and lot-specific purity are provided by the manufacturer and COA.
  • Derived from Salmonella typhosa.
  • White to off-white powder formulation.
  • TLR-4 agonist and immune activator.
  • Used as vaccine adjuvant and immunology reagent.
  • Recommended frozen storage for long-term stability.
  • Lot-specific purity and analytical data available via COA.
Catalog No. 50-002-97134 Supplier Medchemexpress LLC Supplier No. HYD1056B410MG
Lipopolysaccharides from Salmonella typhosa are complex bacterial endotoxins composed of lipid A, a core oligosaccharide, and an O-antigen polysaccharide. Supplied as a white to off-white powder for research use, they act as Toll-like receptor 4 (TLR-4) agonists and can be used as immunostimulatory reagents or vaccine adjuvants. Storage recommendations and lot-specific purity are provided by the manufacturer and COA.
  • Derived from Salmonella typhosa.
  • White to off-white powder formulation.
  • TLR-4 agonist and immune activator.
  • Used as vaccine adjuvant and immunology reagent.
  • Recommended frozen storage for long-term stability.
  • Lot-specific purity and analytical data available via COA.