Learn More
Cusabio Technology LLC lipoprotein lipase
Catalog No. 5000049696
Encompass_Preferred
This kit detects mouse lipoprotein lipase (LPL) in various biological samples including serum plasma cell culture supernates and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 625 ng/mL to 40 ng/mL with a sensitivity of 0 156 ng/mL ensuring precise measurement across physiological concentrations The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Optical density is measured at 450 nm to determine LPL levels making it suitable for researchers studying lipid metabolism cardiovascular diseases or related biomedical pathways requiring reliable quantification of this enzyme in diverse experimental conditions
Catalog No. 50-000-49696 Supplier Cusabio Technology LLC Supplier No. CSBE08495M
This kit detects mouse lipoprotein lipase (LPL) in various biological samples including serum plasma cell culture supernates and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 625 ng/mL to 40 ng/mL with a sensitivity of 0 156 ng/mL ensuring precise measurement across physiological concentrations The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Optical density is measured at 450 nm to determine LPL levels making it suitable for researchers studying lipid metabolism cardiovascular diseases or related biomedical pathways requiring reliable quantification of this enzyme in diverse experimental conditions