Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipoprotein Lipase from Ps50MG
Catalog No. 502903385
Encompass
Lipoprotein Lipase (LPL) is majorly secreted by myocytes and adipocytes in humans and is crucial for triglyceride homeostatis. Mutations in the catalytic domain of LPL impairs its interaction with glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1 (GPIHBP1). The N-terminal catalytic domain is essential for lipolysis. The C-terminal is crucial for binding lipoproteins. Altered LPL levels may play role in the pathogenesis of atherosclerosis coronary heart disease and chronic lymphocytic leukemia.
Catalog No. 50-290-3385 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6233550MG
Lipoprotein Lipase (LPL) is majorly secreted by myocytes and adipocytes in humans and is crucial for triglyceride homeostatis. Mutations in the catalytic domain of LPL impairs its interaction with glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1 (GPIHBP1). The N-terminal catalytic domain is essential for lipolysis. The C-terminal is crucial for binding lipoproteins. Altered LPL levels may play role in the pathogenesis of atherosclerosis coronary heart disease and chronic lymphocytic leukemia.