Learn More
Cell Biolabs Inc Lipoprotein Lipase (LPL) Activity Assay Kit (Fluorometric), 100 assays
Catalog No. NC0521769
Encompass_Preferred
Lipoprotein Lipase (LPL) is the key plasma lipase responsible for the hydrolysis of the triglyceride core in VLDL and chylomicrons. Cell Biolabs' Lipoprotein Lipase Activity Assay provides a convenient assay to quantify LPL activity in plasma, serum, and lysates. LPL activity levels are quantified in a 96-well fluorescence plate reader. This kit also detects endothelial + hepatic lipase activity. Each kit provides sufficient reagents for 100 assays including standards and unknown samples.
Catalog No. NC0521769 Supplier Cell Biolabs Inc Supplier No. STA610
Lipoprotein Lipase (LPL) is the key plasma lipase responsible for the hydrolysis of the triglyceride core in VLDL and chylomicrons. Cell Biolabs' Lipoprotein Lipase Activity Assay provides a convenient assay to quantify LPL activity in plasma, serum, and lysates. LPL activity levels are quantified in a 96-well fluorescence plate reader. This kit also detects endothelial + hepatic lipase activity. Each kit provides sufficient reagents for 100 assays including standards and unknown samples.