Learn More
Kalen Biomedical LLC LIPOROTEIN DEPLETED SERA 100ML
Supplier:  Kalen Biomedical LLC 8801002
Encompass_Preferred
Lipoprotein Depleted Fetal Bovine Serum. The sera are prepared by ultracentrifugation in a potassium bromide solution followed by removal of the fraction with a denisty of less than 1.25g/ml. 100 mL.
Catalog No. NC0009148