Learn More
Sigma Aldrich Fine Chemicals Biosciences LiposoFast Liposome Factor1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z3734001EA
Encompass
Fast efficient formation of uniform-sized unilamellar liposomes. The principle is simple a lipid emulsion is repeatedly extruded through a porous polycarbonate membrane forced back and forth by specially modified gas-tight syringes attached to the membrane support capsule. With the syringes provided unit has 0.5 mL capacity and virtually zero dead volume. Membranes from 100 to 400 nm pore size available.
Catalog No. 50-290-8674