Learn More
ENCAPSULA NANAOSCIENCE LLC LIPOSOMAL CLODRONATE 5ML
Catalog No. NC0337390
Encompass
Clodronate liposomes for macrophage depletion ( 5ml) Suspended in Phosphate Buffered Saline at pH 7.4 •Clodronate encapsulated inside multilamellar liposome aqueous spaces •Extruded through 2 µm membranes to ensure that liposomes easily pass through lung capillaries •Prepared and packaged under sterile conditions
Catalog No. NC0337390 Supplier ENCAPSULA NANAOSCIENCE LLC Supplier No. CLD89095ML
Clodronate liposomes for macrophage depletion ( 5ml) Suspended in Phosphate Buffered Saline at pH 7.4 •Clodronate encapsulated inside multilamellar liposome aqueous spaces •Extruded through 2 µm membranes to ensure that liposomes easily pass through lung capillaries •Prepared and packaged under sterile conditions