Learn More
Kyfora Bio LipoVector Solution 5
Catalog No. 502743939
Encompass_Preferred
Kyfora Bio's LipoVector Solution is a new ready-to-use liposomal transfection reagent designed for the delivery of RNA, DNA, and proteins into eukaryotic cells both in vitro and in vivo. LipoVector can reliably produce recombinant proteins, monoclonal antibodies, and viral vectors in a wide range of cell lines with low cytotoxicity. It supports transient and stable gene expression, co-transfections and is an effective tool for intracellular nucleotide delivery.
Catalog No. 50-274-3939 Supplier Kyfora Bio Supplier No. 145665
Kyfora Bio's LipoVector Solution is a new ready-to-use liposomal transfection reagent designed for the delivery of RNA, DNA, and proteins into eukaryotic cells both in vitro and in vivo. LipoVector can reliably produce recombinant proteins, monoclonal antibodies, and viral vectors in a wide range of cell lines with low cytotoxicity. It supports transient and stable gene expression, co-transfections and is an effective tool for intracellular nucleotide delivery.