Learn More
Medchemexpress LLC Lipoxamycin hemisulfate | 11075-87-9 | 100.0% | 50 MG
SDP
Catalog No. 5000233020
Encompass_Preferred
Lipoxamycin hemisulfate is an antifungal antibiotic and a potent serine palmitoyltransferase (SPT) inhibitor used as a research reagent. It is supplied as a high-purity solid suitable for biochemical and cellular studies.
  • Antifungal antibiotic and SPT inhibitor (IC50 = 21 nM).
  • High purity, suitable for research applications.
  • Supplied as a solid for ease of handling and storage.
  • Available in multiple package sizes, including 50 mg.
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-33020 Supplier Medchemexpress LLC Supplier No. HY119759A50MG
Lipoxamycin hemisulfate is an antifungal antibiotic and a potent serine palmitoyltransferase (SPT) inhibitor used as a research reagent. It is supplied as a high-purity solid suitable for biochemical and cellular studies.
  • Antifungal antibiotic and SPT inhibitor (IC50 = 21 nM).
  • High purity, suitable for research applications.
  • Supplied as a solid for ease of handling and storage.
  • Available in multiple package sizes, including 50 mg.
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).