Learn More
Cayman Chemical LIpoxamycInhemIsulfate 10mg
SDP
Catalog No. 502916505
Encompass_Preferred
A serine palmitoyltransferase inhibitor (IC50 = 21 nM for the S. cerevisiae enzyme) and antifungal agent; inhibits human SPT with an IC50 value ~10-fold lower than the S. cerevisiae enzyme; active against several species of Candida and Cryptococcus, as well as S. cerevisiae (MICs = 0.5-16, 0.25-0.5, and 16 UG/ml, respectively)
Catalog No. 50-291-6505 Supplier Cayman Chemical Supplier No. 3774710 MG
A serine palmitoyltransferase inhibitor (IC50 = 21 nM for the S. cerevisiae enzyme) and antifungal agent; inhibits human SPT with an IC50 value ~10-fold lower than the S. cerevisiae enzyme; active against several species of Candida and Cryptococcus, as well as S. cerevisiae (MICs = 0.5-16, 0.25-0.5, and 16 UG/ml, respectively)