Learn More
Cayman Chemical Lipoxin A4d5 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000773725 UG
Encompass_Preferred
An internal standard for the quantification of Lipoxin A4 by GC- and LC-MS
Catalog No. 50-288-9957