Learn More
Cayman Chemical Lipoxin A4d5 GC MS Standards
SDP
Catalog No. 502887796
Encompass_Preferred
An internal standard for the quantification of Lipoxin A4 by GC- and LC-MS
Catalog No. 50-288-7796 Supplier Cayman Chemical Supplier No. 1000773710 UG
An internal standard for the quantification of Lipoxin A4 by GC- and LC-MS