Learn More
Cayman Chemical Lipoxin A5 Fatty Acids and Der
Supplier: Cayman Chemical 10011453100 UG
A 5-series LO derived from EPA; contracts pulmonary parenchymal strips from guinea pig with similar potency to that of LXA4 and LXB4, yet does not exert the vasodilatory effects on aortic smooth muscle exhibited by LXA4 and LXB4