Learn More
Cayman Chemical Lipoxin A5 Fatty Acids and Der
SDP
Catalog No. 502890092
Encompass_Preferred
A 5-series LO derived from EPA; contracts pulmonary parenchymal strips from guinea pig with similar potency to that of LXA4 and LXB4, yet does not exert the vasodilatory effects on aortic smooth muscle exhibited by LXA4 and LXB4
Catalog No. 50-289-0092 Supplier Cayman Chemical Supplier No. 1001145325 UG
A 5-series LO derived from EPA; contracts pulmonary parenchymal strips from guinea pig with similar potency to that of LXA4 and LXB4, yet does not exert the vasodilatory effects on aortic smooth muscle exhibited by LXA4 and LXB4