Learn More
Cayman Chemical Lipoxin B4-AChE Tracer, 100 dtn
SDP
Catalog No. 502942135
Encompass_Preferred
Lipoxin B4-AChE Tracer has been tested and formulated to work exclusively with Cayman’s Lipoxin B4 ELISA Kit
Catalog No. 50-294-2135 Supplier Cayman Chemical Supplier No. 401920100 DTN
Lipoxin B4-AChE Tracer has been tested and formulated to work exclusively with Cayman’s Lipoxin B4 ELISA Kit