Learn More
Medchemexpress LLC HY-12726 5mg Medchemexpress, Liproxstatin-1 CAS:950455-15-9 Purity:>98%
SDP
Catalog No. 502028911
Encompass_Preferred
Medchemexpress, HY-12726 5mg Liproxstatin-1 CAS:950455-15-9 Liproxstatin-1 is a potent ferroptosis inhibitor and inhibits ferroptotic cell death (IC50=22 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8911 Supplier Medchemexpress LLC Supplier No. HY127265MG
Add to Cart
Add to Cart
Medchemexpress, HY-12726 5mg Liproxstatin-1 CAS:950455-15-9 Liproxstatin-1 is a potent ferroptosis inhibitor and inhibits ferroptotic cell death (IC50=22 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.