Learn More
Kenlor Industries Inc Kenlor Dropper Urine Control Level 1 & Level 2 - 3 X 10 mL each
Catalog No. NC0108721
Encompass_Preferred
Kenlor semi-quantitative dipstick liquid urine control has values for Leukocytes, Urobilinogen, Bilirubin, Glucose, Blood, pH, Nitrite, Ketones, Protein, & Specific Gravity, plus HCG, Creatine, Microalbumin. Stable for 2 years at 2-8°C, 30 days at room temperature. Stable for 30 days at room temperature or 180 days at 2-8°C after opening.
Catalog No. NC0108721 Supplier Kenlor Industries Inc Supplier No. 210306
Kenlor semi-quantitative dipstick liquid urine control has values for Leukocytes, Urobilinogen, Bilirubin, Glucose, Blood, pH, Nitrite, Ketones, Protein, & Specific Gravity, plus HCG, Creatine, Microalbumin. Stable for 2 years at 2-8°C, 30 days at room temperature. Stable for 30 days at room temperature or 180 days at 2-8°C after opening.