Learn More
ARCUS ABSORBENTS INC Liquid Waste Solidifier, 40 lb bag
Catalog No. NC1091903
Encompass_Preferred
LWS is a dry powder used to solidify and encapsulate water based spills.
Catalog No. NC1091903 Supplier ARCUS ABSORBENTS INC Supplier No. SLWS40
LWS is a dry powder used to solidify and encapsulate water based spills.