Learn More
Taylor Wharton LIQ.WITH.DEVICE 25LD/35LD/50LD
Catalog No. NC9959429
Encompass_Preferred
The liquid withdrawal device enables the LD25, LD35, or LD50 Dewar's to conveniently pressure transfer liquid nitrogen at low pressure to assure safe dispensing through a phase separator or an accessory transfer hose.
Catalog No. NC9959429 Supplier Taylor Wharton Supplier No. D0508C03
The liquid withdrawal device enables the LD25, LD35, or LD50 Dewar's to conveniently pressure transfer liquid nitrogen at low pressure to assure safe dispensing through a phase separator or an accessory transfer hose.