Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KOVA™ Liqua-Trol™ Liquid Controls
GSA_VA

Catalog No. 22749759 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Type:
Liqua-Trol Liquid Control with HCG
Liqua-Trol Liquid Control with Microscopics
Liqua-Trol Liquid Control with Microscopics and HCG
Level:
Leve I (Abnormal) and II (Normal)
Level I (Abnormal)
Level II (Normal)
Quantity:
6 x 15 mL
2 x 120 mL
4 x 120 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Level Quantity
22-749-759 Liqua-Trol Liquid Control with Microscopics Level I (Abnormal) 4 x 120 mL
22-749-755 Liqua-Trol Liquid Control with HCG Leve I (Abnormal) and II (Normal) 6 x 15 mL
22-749-758 Liqua-Trol Liquid Control with Microscopics Level I (Abnormal) 2 x 120 mL
22-749-757 Liqua-Trol Liquid Control with Microscopics and HCG Level II (Normal) 4 x 120 mL
22-749-756 Liqua-Trol Liquid Control with Microscopics and HCG Level II (Normal) 2 x 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-749-759 Supplier KOVA™ Supplier No. K87177
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use bi-level liquid control for use with all major brands of urine chemistry dipsticks

 

KOVA Liqua-Trol l control helps to validate the performance of urine chemistry test strips and readers and to focus on microscopic sediment analysis. Choose the KOVA controls that best meet the needs of  your lab. For ready-to-use Urinalysis Controls, choose bi-level liquid control for all major brands of urine chemistry dipsticks.


  • Ready-to-use liquid human urine control
  • External quality control of physical, chemical and microscopic examination of urine specimens
  • Available with or without microscopics
  • Three levels of controls  to monitor complete decision range for urine strip chemistries
  • Value assignments available on all major systems for visual and instrument analysis
  • Stability: stable until the expiration date stated on the label, when stored between 2° and 8°C. Do not freeze. Liqua-Trol can be stored at room temperature (20° to 25°C) for up to 30 days. Controls used at room temperature are for reagent strip and hCG testing only. Label the bottle with the date it was originally brought to room temperature to ensure proper performance.

Recommended Storage

Stable until expiration date stated on the label, when stored between 2° to 8°C. Do not freeze. Liqua-Trol can be stored at room temperature (20° to 25°C) for up to 30 days.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CLSI and CLIA, ANSI Z400.5-2004
Color Yellow, Green or Amber
Content And Storage 2°C to 8°C
Description Liqua-Trol Liquid Control with microscopics
For Use With (Equipment) Urine chemistry dipsticks
Type Liqua-Trol Liquid Control with Microscopics
Intended Use Validate performance of urine chemistry test strips and readers and to focus on microscopic sediment analysis
Level Level I (Abnormal)
Quantity 4 x 120 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.