Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liquefied Phenol, Spectrum™ Chemical
SDP

Catalog No. 18621284 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-284 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-621-284 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P15922.5LTPV

missing translation for 'validMainframeMsg'

Specifications

Quantity 2.5 L
Specific Gravity 1.05
Physical Form Liquid
For Use With (Application) Disinfectant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.