Learn More
Metal Working Chemical & Equipment Co LIQUI CULT FUEL TEST KIT 10/PK
Catalog No. NC9584242
Encompass
Test kits are used to detect microbial growth in fluids in the laboratory and in the workplace. Areas of application include water, metalworking fluids, hydrocarbons, and other liquids such as cleaners, cosmetics, liquid soaps and detergents, etc.
Catalog No. NC9584242 Supplier Metal Working Chemical & Equipment Co Supplier No. LC100
Test kits are used to detect microbial growth in fluids in the laboratory and in the workplace. Areas of application include water, metalworking fluids, hydrocarbons, and other liquids such as cleaners, cosmetics, liquid soaps and detergents, etc.