Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liqui-Gel 29:1, MP Biomedicals

Catalog No. ICN800803 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN800803 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN800803 Supplier MP Biomedicals, Inc Supplier No. 04800803
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 40% (w/v) solution of Ultra Pure Acrylamide (38.67%) and bis-Acrylamide (1.33%) in specially-prepared deionized water. Final ratio is 29:1

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Liqui-Gel™ 29:1
Quantity 500 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.