Learn More
Amsco Scientific Steris LIQUI-JET 2, 4 X 1GAL
Catalog No. NC1872646
Encompass_Preferred
Liqui-Jet® 2 Instrument Detergent. Safe on aluminum/anodized aluminum surgical instruments and aluminum containersContains sequestering agents which makes the detergent effective for a variety of water types, including hard waterLow-foaming formula can be used in ultrasonic and automated washers & washer/disinfector units
This item is not returnable. View return policy
Catalog No. NC1872646 Supplier Amsco Scientific Steris Supplier No. 103708
Liqui-Jet® 2 Instrument Detergent. Safe on aluminum/anodized aluminum surgical instruments and aluminum containersContains sequestering agents which makes the detergent effective for a variety of water types, including hard waterLow-foaming formula can be used in ultrasonic and automated washers & washer/disinfector units