Learn More
Andwin Scientific LIQUI-NOX 1 GALLIQUI-NOX 1 G
SDP
Catalog No. 50997501
Encompass_Preferred
The Liqui-Nox is a specialized cleaning detergent for laboratory equipments. With dilution ratio of 1:100, this safe, odorless, biodegradable, and concentrated cleaning product can be rinsed without any interfering residue. This product is completely soluble in either hard or soft water. It is a basic cleaner with a pH of 8.5. This is sold in a quantity of 1 gallon.
Catalog No. 50-997-501 Supplier Andwin Scientific Supplier No. 60517
The Liqui-Nox is a specialized cleaning detergent for laboratory equipments. With dilution ratio of 1:100, this safe, odorless, biodegradable, and concentrated cleaning product can be rinsed without any interfering residue. This product is completely soluble in either hard or soft water. It is a basic cleaner with a pH of 8.5. This is sold in a quantity of 1 gallon.