Learn More
Andwin Scientific LIQUI-NOX DETERGENT 1QT 12/CS
SDP
Catalog No. NC9302754
Encompass_Preferred
The Liquinox is ideal for use in sanitizing analytical and EPA sampling equipments. This is completely soluble in both hot or cold and hard or soft water. It does not leave any residue with its special free-rinsing formula. This is free of phosphate and is noncorrosive. It is sold in a case of 12.
Catalog No. NC9302754 Supplier Andwin Scientific Supplier No. 21837005
The Liquinox is ideal for use in sanitizing analytical and EPA sampling equipments. This is completely soluble in both hot or cold and hard or soft water. It does not leave any residue with its special free-rinsing formula. This is free of phosphate and is noncorrosive. It is sold in a case of 12.