Learn More
Andwin Scientific LIQUI-NOX DETERGENT 1QT
SDP
Catalog No. NC0355924
Encompass_Preferred
The Liqui-Nox is a specialized cleaning detergent for laboratory equipments. With dilution ratio of 1:100, this safe, odorless, biodegradable, and concentrated cleaning product can be rinsed without any interfering residue. This product is completely soluble in either hard or soft water. It is a basic cleaner with a pH of 8.5. This is sold in a quantity of 1 quart.
Catalog No. NC0355924 Supplier Andwin Scientific Supplier No. 21837005EA
The Liqui-Nox is a specialized cleaning detergent for laboratory equipments. With dilution ratio of 1:100, this safe, odorless, biodegradable, and concentrated cleaning product can be rinsed without any interfering residue. This product is completely soluble in either hard or soft water. It is a basic cleaner with a pH of 8.5. This is sold in a quantity of 1 quart.