Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Liquid Absorbing Sheets
SDP

Catalog No. 22446240 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
4.25 x 2.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-240 4.25 x 2.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-446-240 Supplier Mopec Supplier No. FS010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features an immobilizing agent that reacts on contact with blood, urine, and other body fluids for effective fluid containment.

  • Absorbs from a few drops up to 200 mL of liquid.
  • Provides greater absorption capacity than standard liquid absorbing packets.
  • Immobilizing agent gels body fluid on contact
  • Collects blood and urine

Specifications

Dimensions (L x W) 4.25 x 2.25 in.
Product Type Liquid Absorbing Sheet
Color Transparent
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.