Learn More
Grainger Auralex Liquid Adhesive, Urethane, Water-Based, 10.2 oz
Catalog No. 502782471
Encompass_Preferred
Tubetak Pro™ is a super-strong, urethane-based liquid adhesive designed for the permanent mounting of foam or diffusors. Applied via a standard caulking gun, this water-based adhesive provides reliable, long-lasting durability for wall and ceiling installations.
  • Provides a strong, permanent bond that is heat-resistant, impact-resistant, and waterproof.
  • Features low VOC content.
  • Guaranteed not to oxidize foam products.
  • Covers up to 32 sq. ft. per 10.2 oz tube.
  • Suitable for air shipping.
Catalog No. 50-278-2471 Supplier Grainger Supplier No. 19MP44
Tubetak Pro™ is a super-strong, urethane-based liquid adhesive designed for the permanent mounting of foam or diffusors. Applied via a standard caulking gun, this water-based adhesive provides reliable, long-lasting durability for wall and ceiling installations.
  • Provides a strong, permanent bond that is heat-resistant, impact-resistant, and waterproof.
  • Features low VOC content.
  • Guaranteed not to oxidize foam products.
  • Covers up to 32 sq. ft. per 10.2 oz tube.
  • Suitable for air shipping.