Learn More
Perkin Elmer US LLC Liquid and Mull Complete Sampling Kit
Catalog No. 502117207
Encompass_Preferred
PerkinElmer provides FT-IR Sampling Kits to help the newcomer to FT-IR spectroscopy choose the more frequently required items, and to simplify ordering and storage. KBr has been chosen for the transmitting material.
Catalog No. 50-211-7207 Supplier Perkin Elmer US LLC Supplier No. L1365311
PerkinElmer provides FT-IR Sampling Kits to help the newcomer to FT-IR spectroscopy choose the more frequently required items, and to simplify ordering and storage. KBr has been chosen for the transmitting material.