Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Science First™ Liquid Conductivity Indicator
SDPSCI_ED

Catalog No. p-2366384
Change view
Click to view available options
Type:
Conductivity Indicator
Replacement Bulb
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S43478 Conductivity Indicator
S43478BLB Replacement Bulb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S43478 Supplier Science First™ Supplier No. 6112260
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Requires

Light bulb, 7.5w

Determine the conductivity of different liquids or concentrations.

  • Qualitative device causes a 7.5w light bulb to glow when water conducts electricity
  • Cord on the side cannot contact water, eliminating shock hazards
  • Plastic molded body with recessed electrical connections and electrodes
  • Attaches to standard ring stand (up to 0.5 in. dia. [12.7mm]) with thumbscrew clamp (sold separately)
  • Comes with illustrated instructions
  • Weight: 3.88 oz. (0.11kg)

Operates on 110V, AC only

Specifications

For Use With (Application) Determine the conductivity of liquids or concentrations
Type Conductivity Indicator
Voltage 110 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.