Learn More
Chemglass Life Sciences Extractor, Liquid-Liquid, Continous, 45/50, 24/40
SDP
Catalog No. CG136401
Encompass_Preferred
Extractor, Liquid-Liquid, Continuous, 45/50, 24/40. Similar in design to CG-1360 but with the addition of a 2mm PTFE stopcock in the return tube. Stopcock stops the flow after the extraction is complete. For samples of 1 liter plus approximately 200mL of extracting solvent. Extractor has a 45/50 top outer joint and a 24/40 inner joint on the uptake tube. Flask and condenser must be ordered separately.
Catalog No. CG136401 Supplier Chemglass Life Sciences Supplier No. CG136401
Extractor, Liquid-Liquid, Continuous, 45/50, 24/40. Similar in design to CG-1360 but with the addition of a 2mm PTFE stopcock in the return tube. Stopcock stops the flow after the extraction is complete. For samples of 1 liter plus approximately 200mL of extracting solvent. Extractor has a 45/50 top outer joint and a 24/40 inner joint on the uptake tube. Flask and condenser must be ordered separately.