Learn More
Roche Diagnostics Stains Liquid Coverslip, One each
Catalog No. NC9710885 Shop All Roche Diagnostics Products
Encompass
LCS It is Ventana reagent required but not supplied with ChipMap kit
Catalog No. NC9710885 Supplier Roche Diagnostics Supplier No. 05264839001
LCS It is Ventana reagent required but not supplied with ChipMap kit