Learn More
Biolog Inc LDT - Liquid Dental Transport 1mL, 10 Tubes/Pkg
SDP
Catalog No. NC0188292
Encompass
This item has a minimum qty of 2 per supplier requirements.
Liquid Dental Transport Med. (LDT) is a buffered mineral salt based liquid Med.. Designed as a non-nutritive holding Med., it maintains viability of microorganisms through collection & transport of clinical specimens. This media is prepared, dispensed, & packaged under oxygen-free conditions.LDT contains buffered mineral salts in a liquid form w/ sodium thioglycollate & cysteine added to provide a reduced environment. This combination has been prepared to provide an environment which maintains the viability of most microorganisms without significant multiplication & allows for dilution of inhibitors present in clinical material. This Med. is designed to meet the stringent viability requirements of obligate anaerobes.
Catalog No. NC0188292 Supplier Biolog Inc Supplier No. AS916
This item has a minimum qty of 2 per supplier requirements.
Liquid Dental Transport Med. (LDT) is a buffered mineral salt based liquid Med.. Designed as a non-nutritive holding Med., it maintains viability of microorganisms through collection & transport of clinical specimens. This media is prepared, dispensed, & packaged under oxygen-free conditions.LDT contains buffered mineral salts in a liquid form w/ sodium thioglycollate & cysteine added to provide a reduced environment. This combination has been prepared to provide an environment which maintains the viability of most microorganisms without significant multiplication & allows for dilution of inhibitors present in clinical material. This Med. is designed to meet the stringent viability requirements of obligate anaerobes.