Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ashton Pumpmatic Liquid Dispensing System
SDP

Catalog No. 15118002
Encompass
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
15-118-002 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-118-002 Supplier Ashton Pumpmatic Supplier No. 1110
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Delivers accurate volumes without drips or spills with individually wrapped

  • Rapid, drop-free fluid sampling, transferring, and dispensing
  • Speeds and simplifies fluid sampling from 1 mL to 10 mL per sample
  • Serological pipette/dispenser combination offers accuracy of a reusable at the cost of a disposable
  • Controlled rate of sampling and dispensing delivers accurate volumes without drips or spills
  • Hazardous liquids do not need to be closer than arm's length
  • Transparent, totally closed system
  • Features accurate, clearly marked, easily-read graduations
  • Smooth-action syringe and plunger with Luer Lok™ permit rapid, one-handed sampling and dispensing
  • Lightweight, unbreakable, clear polystyrene

Specifications

Autoclavable Not Autoclavable
Material Polystyrene, Polypropylene
Sterility Sterile
Graduated Yes
Packaging Type Individually Wrapped
Color Clear
Volume (Metric) 10 mL
Type Serological Pipet, Single-use
Disposable Single-use
Fit for Purpose Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.