Learn More
Grainger Liquid Filled Food Service Thermometer
Catalog No. 502750738
Encompass_Preferred
The Taylor 14F311 mechanical food service thermometer is a dial thermometer designed for commercial kitchen use. It has a stainless steel stem that is 5.5 inches long with a 1.5 inch dial face. The dial is color coded to indicate temperature zones from 0-220°F/-18-104°C. It can be calibrated and has a pointed tip for easy insertion into food. The dial is encased in a stainless steel case with a hanging hole for storage. This NSF certified thermometer is ideal for monitoring cooking and holding temperatures of food in restaurants and commercial kitchens.
Catalog No. 50-275-0738 Supplier Grainger Supplier No. 14F312
The Taylor 14F311 mechanical food service thermometer is a dial thermometer designed for commercial kitchen use. It has a stainless steel stem that is 5.5 inches long with a 1.5 inch dial face. The dial is color coded to indicate temperature zones from 0-220°F/-18-104°C. It can be calibrated and has a pointed tip for easy insertion into food. The dial is encased in a stainless steel case with a hanging hole for storage. This NSF certified thermometer is ideal for monitoring cooking and holding temperatures of food in restaurants and commercial kitchens.