Learn More
Electron Microscopy Sciences Disposable Liquid-Holding Slide Wells 0.50 thick
SDP
Catalog No. 5034532
Encompass_Preferred
These disposable liquid-holding wells are pre-cut, twin adhesive well stickers (1cm diameter) firmly adhere to microscope slides. Examine specimens with or without cover slips.
Multiple Applications Examine or count tissue culture cells, yeast, bacteria and other  microorganisms.  Piggy-back with Microscope Slide-Grids to facilitate counting and sizing.
Versatile - Compatible with different styles of microscopes (conventional, inverted or dissecting).
Chemically Inert - Made of polyester with an acrylic adhesive. Microscope Slide-Wells are inert and stable under most laboratory conditions.
Thickness: 0.50 mm
Catalog No. 50-345-32 Supplier Electron Microscopy Sciences Supplier No. 7036613
These disposable liquid-holding wells are pre-cut, twin adhesive well stickers (1cm diameter) firmly adhere to microscope slides. Examine specimens with or without cover slips.
Multiple Applications Examine or count tissue culture cells, yeast, bacteria and other  microorganisms.  Piggy-back with Microscope Slide-Grids to facilitate counting and sizing.
Versatile - Compatible with different styles of microscopes (conventional, inverted or dissecting).
Chemically Inert - Made of polyester with an acrylic adhesive. Microscope Slide-Wells are inert and stable under most laboratory conditions.
Thickness: 0.50 mm